Assay Details
Gene Symbol: LOC4333983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777179.2 | XP_015632665.1 | 5 - 6 | 905 | 73 | |
GenBank mRNA | AK068068.1 | - | 5 - 6 | 903 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
