Assay Details
| Gene Symbol: LOC4334089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772606.2 | XP_015628092.1 | 14 - 15 | 1713 | 78 | |
| GenBank mRNA | AK101069.1 | - | 14 - 15 | 1692 | 78 | |
| GQ982584.1 | - | 14 - 15 | 1498 | 78 | ||
More Information
Set Membership: |
|

LOC4334089
Genomic Map