Assay Details
| Gene Symbol: LOC4334089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772606.2 | XP_015628092.1 | 4 - 5 | 719 | 87 | |
| GenBank mRNA | AK101069.1 | - | 4 - 5 | 698 | 87 | |
| GQ982584.1 | - | 4 - 5 | 504 | 87 | ||
More Information
Set Membership: |
|

LOC4334089
Genomic Map