Assay Details
| Gene Symbol: LOC4334089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772606.2 | XP_015628092.1 | 6 - 7 | 978 | 94 | |
| GenBank mRNA | AK101069.1 | - | 6 - 7 | 957 | 94 | |
| GQ982584.1 | - | 6 - 7 | 763 | 94 | ||
More Information
Set Membership: |
|

LOC4334089
Genomic Map