Assay Details
| Gene Symbol: LOC4334116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776761.2 | XP_015632247.1 | 6 - 7 | 1135 | 64 | |
| XM_015776762.2 | XP_015632248.1 | 6 - 7 | 1277 | 64 | ||
| GenBank mRNA | AK073110.1 | - | 6 - 7 | 1277 | 64 | |
More Information
Set Membership: |
|

LOC4334116
Genomic Map