Assay Details
Gene Symbol: LOC4334513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775065.2 | XP_015630551.1 | 1 - 2 | 499 | 65 | |
GenBank mRNA | AK060962.1 | - | 1 - 2 | 507 | 65 | |
AK070287.1 | - | 1 - 2 | 405 | 65 | ||
KX434728.1 | - | 1 - 2 | 254 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
