Assay Details
| Gene Symbol: LOC4334513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775065.2 | XP_015630551.1 | 2 - 3 | 583 | 67 | |
| GenBank mRNA | AK060962.1 | - | 2 - 3 | 591 | 67 | |
| AK070287.1 | - | 2 - 3 | 489 | 67 | ||
| KX434728.1 | - | 2 - 3 | 338 | 67 | ||
More Information
Set Membership: |
|

LOC4334513
Genomic Map