Assay Details
Gene Symbol: LOC4334700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774271.2 | XP_015629757.1 | 10 - 11 | 1025 | 97 | |
XM_015774273.2 | XP_015629759.1 | 10 - 11 | 1026 | 97 | ||
GenBank mRNA | AK070689.1 | - | 4 - 5 | 326 | 97 | |
AK102903.1 | - | 10 - 11 | 963 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
