Assay Details
Gene Symbol: LOC4334700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774271.2 | XP_015629757.1 | 13 - 14 | 1453 | 73 | |
XM_015774273.2 | XP_015629759.1 | 13 - 14 | 1454 | 73 | ||
GenBank mRNA | AK070689.1 | - | 7 - 8 | 754 | 73 | |
AK102903.1 | - | 13 - 14 | 1391 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
