Assay Details
Gene Symbol: LOC4334700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774271.2 | XP_015629757.1 | 15 - 16 | 1623 | 85 | |
XM_015774273.2 | XP_015629759.1 | 15 - 16 | 1624 | 85 | ||
GenBank mRNA | AK070689.1 | - | 9 - 10 | 924 | 85 | |
AK102903.1 | - | 14 - 15 | 1966 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
