Assay Details
Gene Symbol: LOC4334700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774271.2 | XP_015629757.1 | 16 - 17 | 1714 | 69 | |
GenBank mRNA | AK070689.1 | - | 10 - 11 | 1015 | 69 | |
AK102903.1 | - | 15 - 16 | 2057 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
