Assay Details
Gene Symbol: LOC4334700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774271.2 | XP_015629757.1 | 1 - 2 | 266 | 69 | |
XM_015774273.2 | XP_015629759.1 | 1 - 2 | 267 | 69 | ||
GenBank mRNA | AK102903.1 | - | 1 - 2 | 204 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
