Assay Details
Gene Symbol: LOC4334700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774271.2 | XP_015629757.1 | 2 - 3 | 360 | 68 | |
XM_015774273.2 | XP_015629759.1 | 2 - 3 | 361 | 68 | ||
GenBank mRNA | AK102903.1 | - | 2 - 3 | 298 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
