Assay Details
Gene Symbol: LOC4334700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774271.2 | XP_015629757.1 | 5 - 6 | 633 | 138 | |
XM_015774273.2 | XP_015629759.1 | 5 - 6 | 634 | 138 | ||
GenBank mRNA | AK102903.1 | - | 5 - 6 | 571 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
