Assay Details
Gene Symbol: LOC4334700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774271.2 | XP_015629757.1 | 7 - 8 | 793 | 83 | |
XM_015774273.2 | XP_015629759.1 | 7 - 8 | 794 | 83 | ||
GenBank mRNA | AK070689.1 | - | 1 - 2 | 94 | 83 | |
AK102903.1 | - | 7 - 8 | 731 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
