Assay Details
Gene Symbol: LOC4334700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774271.2 | XP_015629757.1 | 9 - 10 | 936 | 67 | |
XM_015774273.2 | XP_015629759.1 | 9 - 10 | 937 | 67 | ||
GenBank mRNA | AK070689.1 | - | 3 - 4 | 237 | 67 | |
AK102903.1 | - | 9 - 10 | 874 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
