Assay Details
| Gene Symbol: LOC4334702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774275.2 | XP_015629761.1 | 1 - 2 | 184 | 78 | |
| GenBank mRNA | AK063598.1 | - | 1 - 2 | 122 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015774275.2 transcripts.
Set Membership: |
|

LOC4334702
Genomic Map