Assay Details
Gene Symbol: LOC4334806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772767.2 | XP_015628253.1 | 3 - 4 | 767 | 109 | |
GenBank mRNA | AK066656.1 | - | 2 - 3 | 745 | 109 | |
AK104812.1 | - | 2 - 3 | 673 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
