Assay Details
Gene Symbol: LOC4334806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772767.2 | XP_015628253.1 | 4 - 5 | 900 | 75 | |
GenBank mRNA | AK066656.1 | - | 3 - 4 | 878 | 75 | |
AK104812.1 | - | 3 - 4 | 806 | 75 | ||
Y10253.1 | - | 3 - 4 | 893 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
