Assay Details
Gene Symbol: LOC4334857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776770.2 | XP_015632256.1 | 5 - 6 | 1040 | 101 | |
XM_015776771.2 | XP_015632257.1 | 5 - 6 | 1036 | 101 | ||
GenBank mRNA | AK073135.1 | - | 5 - 6 | 983 | 101 | |
AK105297.1 | - | 5 - 6 | 972 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
