Assay Details
Gene Symbol: LOC4335391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778396.2 | XP_015633882.1 | 1 - 2 | 1710 | 72 | |
GenBank mRNA | AK062593.1 | - | 1 - 2 | 1471 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
