Assay Details
Gene Symbol: LOC4336155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015779501.2 | XP_015634987.1 | 2 - 3 | 2795 | 99 | |
XM_015779502.2 | XP_015634988.1 | 2 - 3 | 3568 | 99 | ||
XM_015779503.2 | XP_015634989.1 | 2 - 3 | 2777 | 99 | ||
GenBank mRNA | AK065827.1 | - | 2 - 3 | 2714 | 99 | |
AK101478.1 | - | 2 - 3 | 2779 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
