Assay Details
| Gene Symbol: LOC4336389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781208.1 | XP_015636694.1 | 2 - 3 | 376 | 75 | |
| XM_015781209.1 | XP_015636695.1 | 2 - 3 | 376 | 75 | ||
| XM_015781210.1 | XP_015636696.1 | 2 - 3 | 376 | 75 | ||
| GenBank mRNA | AK066850.1 | - | 2 - 3 | 272 | 75 | |
| AK068772.1 | - | 2 - 3 | 272 | 75 | ||
More Information
Set Membership: |
|

LOC4336389
Genomic Map