Assay Details
Gene Symbol: LOC4336457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778598.2 | XP_015634084.1 | 1 - 2 | 339 | 68 | |
GenBank mRNA | AK070324.1 | - | 1 - 2 | 315 | 68 | |
AK099545.1 | - | 1 - 2 | 316 | 68 | ||
AK104433.1 | - | 1 - 2 | 315 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
