Assay Details
Gene Symbol: LOC9268164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778879.2 | XP_015634365.1 | 1 - 2 | 1560 | 65 | |
GenBank mRNA | AK099968.1 | - | 1 - 2 | 1405 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
