Assay Details
Gene Symbol: LOC9268164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778879.2 | XP_015634365.1 | 4 - 5 | 2426 | 78 | |
GenBank mRNA | AK099968.1 | - | 3 - 4 | 2362 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
