Assay Details
| Gene Symbol: LOC9268618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781419.2 | XP_015636905.1 | 4 - 5 | 541 | 106 | |
| XM_015781422.2 | XP_015636908.1 | 4 - 5 | 541 | 106 | ||
| GenBank mRNA | AK242596.1 | - | 4750 | 106 | ||
| AK288235.1 | - | 4752 | 106 | |||
More Information
Set Membership: |
|

LOC9268618
Genomic Map