Assay Details
| Gene Symbol: LOC4337748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783064.2 | XP_015638550.1 | 1 | 489 | 66 | |
| GenBank mRNA | AK120814.1 | - | 1 | 257 | 66 | |
More Information
Set Membership: |
|

LOC4337748
Genomic Map