Assay Details
| Gene Symbol: LOC4338161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785017.2 | XP_015640503.1 | 5 - 6 | 792 | 120 | |
| GenBank mRNA | AK067996.1 | - | 4 - 5 | 538 | 120 | |
| HQ013294.1 | - | 4 - 5 | 549 | 120 | ||
More Information
Set Membership: |
|

LOC4338161
Genomic Map