Assay Details
| Gene Symbol: LOC4338161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785017.2 | XP_015640503.1 | 8 - 9 | 1083 | 154 | |
| GenBank mRNA | AK067996.1 | - | 7 - 8 | 829 | 154 | |
| HQ013294.1 | - | 7 - 8 | 840 | 154 | ||
More Information
Set Membership: |
|

LOC4338161
Genomic Map