Assay Details
| Gene Symbol: LOC4338940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784881.2 | XP_015640367.1 | 18 - 19 | 5634 | 82 | |
| XM_015784882.2 | XP_015640368.1 | 18 - 19 | 5631 | 82 | ||
| GenBank mRNA | AK103871.1 | - | 12 - 13 | 1255 | 82 | |
| FJ536320.1 | - | 17 - 18 | 5271 | 82 | ||
More Information
Set Membership: |
|

LOC4338940
Genomic Map