Assay Details
Gene Symbol: LOC4338944 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015783899.2 | XP_015639385.1 | 9 - 10 | 4554 | 84 | |
XM_015783900.2 | XP_015639386.1 | 9 - 10 | 4551 | 84 | ||
XM_015783901.2 | XP_015639387.1 | 9 - 10 | 4558 | 84 | ||
XM_015783902.2 | XP_015639388.1 | 9 - 10 | 4558 | 84 |
Gene Symbol: LOC4338940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784881.2 | XP_015640367.1 | 9 - 10 | 4585 | 84 | |
XM_015784882.2 | XP_015640368.1 | 9 - 10 | 4582 | 84 | ||
XM_015784883.2 | XP_015640369.1 | 9 - 10 | 4591 | 84 | ||
XM_015784884.2 | XP_015640370.1 | 9 - 10 | 4594 | 84 | ||
GenBank mRNA | AK103871.1 | - | 3 - 4 | 206 | 84 | |
FJ536320.1 | - | 8 - 9 | 4222 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
