Assay Details
Gene Symbol: LOC4338944 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015783899.2 | XP_015639385.1 | 10 - 11 | 4623 | 89 | |
XM_015783900.2 | XP_015639386.1 | 10 - 11 | 4620 | 89 | ||
XM_015783901.2 | XP_015639387.1 | 10 - 11 | 4627 | 89 | ||
XM_015783902.2 | XP_015639388.1 | 10 - 11 | 4627 | 89 |
Gene Symbol: LOC4338940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784881.2 | XP_015640367.1 | 10 - 11 | 4654 | 89 | |
XM_015784882.2 | XP_015640368.1 | 10 - 11 | 4651 | 89 | ||
XM_015784883.2 | XP_015640369.1 | 10 - 11 | 4660 | 89 | ||
XM_015784884.2 | XP_015640370.1 | 10 - 11 | 4663 | 89 | ||
GenBank mRNA | AK103871.1 | - | 4 - 5 | 275 | 89 | |
FJ536320.1 | - | 9 - 10 | 4291 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
