Assay Details
| Gene Symbol: LOC4338944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783899.2 | XP_015639385.1 | 11 - 12 | 4664 | 118 | |
| XM_015783900.2 | XP_015639386.1 | 11 - 12 | 4661 | 118 | ||
| XM_015783901.2 | XP_015639387.1 | 11 - 12 | 4668 | 118 | ||
| XM_015783902.2 | XP_015639388.1 | 11 - 12 | 4668 | 118 | ||
| Gene Symbol: LOC4338940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784881.2 | XP_015640367.1 | 11 - 12 | 4695 | 118 | |
| XM_015784882.2 | XP_015640368.1 | 11 - 12 | 4692 | 118 | ||
| XM_015784883.2 | XP_015640369.1 | 11 - 12 | 4701 | 118 | ||
| XM_015784884.2 | XP_015640370.1 | 11 - 12 | 4704 | 118 | ||
| GenBank mRNA | AK103871.1 | - | 5 - 6 | 316 | 118 | |
| FJ536320.1 | - | 10 - 11 | 4332 | 118 | ||
More Information
Set Membership: |
|

LOC4338944
Genomic Map