Assay Details
Gene Symbol: LOC4339151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784927.2 | XP_015640413.1 | 3 - 4 | 403 | 88 | |
XM_015784928.2 | XP_015640414.1 | 2 - 3 | 346 | 88 | ||
XM_015784929.2 | XP_015640415.1 | 3 - 4 | 401 | 88 | ||
XM_015784930.2 | XP_015640416.1 | 3 - 4 | 403 | 88 | ||
XM_015784931.2 | XP_015640417.1 | 3 - 4 | 403 | 88 | ||
GenBank mRNA | AK241362.1 | - | 2 - 3 | 346 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
