Assay Details
Gene Symbol: LOC4339663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784356.2 | XP_015639842.1 | 1 - 2 | 252 | 165 | |
GenBank mRNA | AK111749.1 | - | 1 - 2 | 259 | 165 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784356.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
