Assay Details
Gene Symbol: LOC4339663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784356.2 | XP_015639842.1 | 4 - 5 | 3130 | 99 | |
XM_015784357.2 | XP_015639843.1 | 4 - 5 | 3119 | 99 | ||
GenBank mRNA | AK111749.1 | - | 4 - 5 | 3137 | 99 | |
AK120171.1 | - | 3 - 4 | 1464 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
