Assay Details
Gene Symbol: LOC4339663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784356.2 | XP_015639842.1 | 6 - 7 | 3595 | 137 | |
XM_015784357.2 | XP_015639843.1 | 6 - 7 | 3584 | 137 | ||
GenBank mRNA | AK111749.1 | - | 6 - 7 | 3602 | 137 | |
AK120171.1 | - | 5 - 6 | 1929 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
