Assay Details
| Gene Symbol: LOC4339859 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785377.2 | XP_015640863.1 | 16 - 17 | 1497 | 75 | |
| XM_015785378.2 | XP_015640864.1 | 16 - 17 | 1588 | 75 | ||
| GenBank mRNA | AK071985.1 | - | 16 - 17 | 1438 | 75 | |
More Information
Set Membership: |
|

LOC4339859
Genomic Map