Assay Details
| Gene Symbol: LOC4339859 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785377.2 | XP_015640863.1 | 2 - 3 | 229 | 72 | |
| XM_015785378.2 | XP_015640864.1 | 2 - 3 | 320 | 72 | ||
| GenBank mRNA | AK071985.1 | - | 2 - 3 | 170 | 72 | |
| AK103793.1 | - | 2 - 3 | 189 | 72 | ||
More Information
Set Membership: |
|

LOC4339859
Genomic Map