Assay Details
Gene Symbol: LOC4340018 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789000.2 | XP_015644486.1 | 4 - 5 | 1927 | 80 | |
XM_015789001.2 | XP_015644487.1 | 5 - 6 | 804 | 80 | ||
XM_015789002.2 | XP_015644488.1 | 5 - 6 | 798 | 80 | ||
XM_015789003.2 | XP_015644489.1 | 5 - 6 | 736 | 80 | ||
XM_015789004.2 | XP_015644490.1 | 4 - 5 | 1927 | 80 | ||
XM_015789005.2 | XP_015644491.1 | 5 - 6 | 798 | 80 | ||
XM_015789006.2 | XP_015644492.1 | 5 - 6 | 734 | 80 | ||
GenBank mRNA | AB066093.1 | - | 4 - 5 | 635 | 80 | |
AB066094.1 | - | 4 - 5 | 635 | 80 | ||
AF515481.1 | - | 4 - 5 | 611 | 80 | ||
AF515482.1 | - | 4 - 5 | 611 | 80 | ||
AF515483.1 | - | 4 - 5 | 611 | 80 | ||
AK070431.1 | - | 5 - 6 | 782 | 80 | ||
EU735072.1 | - | 4 - 5 | 611 | 80 | ||
KC332294.1 | - | 4 - 5 | 611 | 80 | ||
KT199369.1 | - | 5 - 6 | 681 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
