Assay Details
| Gene Symbol: LOC4340110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788577.2 | XP_015644063.1 | 1 - 2 | 282 | 71 | |
| XM_015788578.2 | XP_015644064.1 | 1 - 2 | 282 | 71 | ||
| XM_015788579.2 | XP_015644065.1 | 1 - 2 | 282 | 71 | ||
| XM_015788580.2 | XP_015644066.1 | 1 - 2 | 282 | 71 | ||
| XM_015788581.2 | XP_015644067.1 | 1 - 2 | 282 | 71 | ||
| XM_015788582.2 | XP_015644068.1 | 1 - 2 | 282 | 71 | ||
| GenBank mRNA | AK120452.1 | - | 1 - 2 | 271 | 71 | |
More Information
Set Membership: |
|

LOC4340110
Genomic Map