Assay Details
| Gene Symbol: LOC4340110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788577.2 | XP_015644063.1 | 4 - 5 | 3494 | 72 | |
| XM_015788581.2 | XP_015644067.1 | 4 - 5 | 3494 | 72 | ||
| XM_015788582.2 | XP_015644068.1 | 4 - 5 | 3494 | 72 | ||
| GenBank mRNA | AK120452.1 | - | 4 - 5 | 3483 | 72 | |
More Information
Set Membership: |
|

LOC4340110
Genomic Map