Assay Details
| Gene Symbol: LOC4340375 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785675.2 | XP_015641161.1 | 2 - 3 | 400 | 76 | |
| GenBank mRNA | AK067746.1 | - | 2 - 3 | 387 | 76 | |
More Information
Set Membership: |
|

LOC4340375
Genomic Map