Assay Details
| Gene Symbol: LOC4341496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787679.2 | XP_015643165.1 | 2 - 3 | 209 | 62 | |
| GenBank mRNA | AK058288.1 | - | 1 - 2 | 97 | 62 | |
| AK058302.1 | - | 1 - 2 | 97 | 62 | ||
| AK064989.1 | - | 1 - 2 | 158 | 62 | ||
| AK104381.1 | - | 1 - 2 | 101 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015787679.2 transcripts.
Set Membership: |
|

LOC4341496
Genomic Map