Assay Details
| Gene Symbol: LOC4341496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787679.2 | XP_015643165.1 | 3 - 4 | 891 | 115 | |
| GenBank mRNA | AK058288.1 | - | 2 - 3 | 779 | 115 | |
| AK058302.1 | - | 2 - 3 | 779 | 115 | ||
| AK064989.1 | - | 2 - 3 | 840 | 115 | ||
| AK104381.1 | - | 2 - 3 | 783 | 115 | ||
More Information
Set Membership: |
|

LOC4341496
Genomic Map