Assay Details
| Gene Symbol: LOC4341648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788830.2 | XP_015644316.1 | 11 - 12 | 1520 | 140 | |
| GenBank mRNA | AK101292.1 | - | 11 - 12 | 1526 | 140 | |
| AK101919.1 | - | 11 - 12 | 1493 | 140 | ||
More Information
Set Membership: |
|

LOC4341648
Genomic Map