Assay Details
| Gene Symbol: LOC4341648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788830.2 | XP_015644316.1 | 6 - 7 | 1112 | 85 | |
| XM_015788831.2 | XP_015644317.1 | 6 - 7 | 1107 | 85 | ||
| GenBank mRNA | AK101292.1 | - | 6 - 7 | 1118 | 85 | |
| AK101919.1 | - | 6 - 7 | 1085 | 85 | ||
More Information
Set Membership: |
|

LOC4341648
Genomic Map