Assay Details
| Gene Symbol: LOC4341870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK068614.1 | - | 10 - 11 | 2448 | 64 | |
| AK120488.1 | - | 10 - 11 | 2448 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4341870| Gene Symbol: LOC4341870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK068614.1 | - | 10 - 11 | 2448 | 64 | |
| AK120488.1 | - | 10 - 11 | 2448 | 64 | ||
Set Membership: |
|