Assay Details
Gene Symbol: LOC4342102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788312.2 | XP_015643798.1 | 4 - 5 | 760 | 120 | |
GenBank mRNA | AK073850.1 | - | 4 - 5 | 404 | 120 | |
AK103177.1 | - | 4 - 5 | 404 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
